Serum-free medium, Panserin MDCK
Supplier: PAN-Biotech
PANBP04-71105EA 1270 SGD
PANBP04-71105
Serum-free medium, Panserin MDCK
Cell Culture Media
Panserin MDCK is a complete serum-free ready-to-use medium for the cultivation of MDCK cells (Madin-Darby canine kidney cells, epithelial cells from normal adult female cocker spaniel) in an adherent culture.
  • Stored at 2 to 8 °C