Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium hexafluoroantimonate ≥98%

Supplier: Alfa Aesar
Warning
Synonyms: Potassium hexafluoroantimonate(V)



Formula: F₆KSb
MW: 274,85 g/mol
Boiling Pt: 1505 °C
Melting Pt: 846 °C
MDL Number: MFCD00043080
CAS Number: 16893-92-8
UN: 1549
ADR: 6.1,III

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 50 g ALFA40146.18
Supplier Article Number:: 40146.18
Expand All / Collapse All

SPECIFICATIONS

Melting point 846°
Formula weight 274.84
CAS No. 16893-92-8
Purity 98% min
Physical form Powder
Odour/scent Odourless
Boiling point 1505°
Molecular formula KSbF6
MDL No. MFCD00043080
UN No. 1549
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty