Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium selenate ≥99%

Supplier: Alfa Aesar
Danger



Formula: K₂O₄Se
MW: 221,15 g/mol
Density: 3,07 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00054125
CAS Number: 7790-59-2
EINECS: 232-214-7
UN: 2630
ADR: 6.1,I
Merck Index: 12,07836

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 10 g ALFAA16135.09
Supplier Article Number:: A16135.09
Expand All / Collapse All

SPECIFICATIONS

Formula weight 221.16
CAS No. 7790-59-2
Purity 99%
Odour/scent Odourless
Molecular formula K2SeO4
MDL No. MFCD00054125
UN No. 2630
Merck reference 14,7666
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping