Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium titanate ≥98% (metals basis)

Supplier: Alfa Aesar



Formula: K₂O₃Ti
MW: 174,07 g/mol
Melting Pt: 826 °C
Density: 3,1 g/cm³ (20 °C)
MDL Number: MFCD00058803
CAS Number: 12030-97-6
EINECS: 234-748-6

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 500 g ALFA14138.36
Supplier Article Number:: 14138.36
Glass bottle for solids 100 g ALFA14138.22
Supplier Article Number:: 14138.22
Expand All / Collapse All

SPECIFICATIONS

Melting point 1615°
Formula weight 174.06
CAS No. 12030-97-6
Purity 98%
Physical form -325 Mesh Powder
Odour/scent Odourless
MDL No. MFCD00058803
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty