VWR® PCR Plates, 96-Well, for ABI PRISM 3700 Series
Supplier: VWR
VWR®
VWRI732-1114EA 43.9 SGD
VWRI732-1114
VWR® PCR Plates, 96-Well, for ABI PRISM 3700 Series
Microplates PCR Plates
Inert PP, natural. Designed to fit ABI PRISM 3700 series thermocyclers. Plates feature a smooth rim that allows better contact inside the block to ensure a perfect fit and easy removal, as well as printed alphanumeric labelling for easy well location.
Designed to fit ABI PRISM 3700 series thermocyclers. Plates feature a smooth rim that allows better contact inside the block to ensure a perfect fit and easy removal, as well as printed alphanumeric labelling for easy well location.
Well volume: 200 μl