VWR®, 70% IPA USP and 30% WFI, Sterile
Supplier: Avantor
VWR®
VWRA141-1444EA 47.3 SGD
VWRA141-1444
VWR®, 70% IPA USP and 30% WFI, Sterile
Cleaning Agents Disinfectants
The cleanroom trigger spray contains 70% IPA (USP Grade) and 30% WFI.
The product is 0.2 μm filtered and filled in an enclosed automated filling line in a NEBB certified ISO Class 5 cleanroom.
The trigger spray is pre-installed onto the bottle and ready to use. Each product is double bagged for cleanroom handling and Gamma irradiated and sterility tested to Sterility Assurance Level (SAL) of 10-6 .