VWR®, Compact Universal Laboratory Centrifuge
Supplier: VWR
VWR®
VWRA521-3316EA 2390 SGD
VWRA521-3316
VWR®, Compact Universal Laboratory Centrifuge
Centrifuges Benchtop Centrifuges
This universal laboratory centrifuge delivers sustained accuracy with any load thanks to the ability to shut off at the slightest hint of imbalance. Thoroughly tested, it functions smoothly even at maximum speed. The centrifuge is compatible with multiple rotors (swing and fixed angle), this compact device offers superior design with precision performance.
  • Brushless DC motor for maintenance free long life
  • Microprocessor controlled centrifuge with variable speed and time setting also features last run memory function
  • Centrifuge with choice of different rotors (swing out and fixed angle) suitable for use with various work protocols
  • Imbalance detection and lid lock safety features
  • Auto rotor detection feature
Ordering information: Please select the correct rotor for the required application.