Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hematoxylin, high purity for histology

Supplier: VWR Chemicals
Synonyms: DcH-48HydroxybrasilinNatural Black I

Nuclei stain used for general animal histology. Also used as a pH indicator: goes from red to yellow over a range of 0.0 to 1.0, and light yellow to violet over a range of 5.0 to 6.0.


Formula: C₁₆H₁₄O₆
MW: 302,28 g/mol
Melting Pt: 291 - 293 °C
Storage Temperature: Ambient
MDL Number: MFCD00078111
CAS Number: 517-28-2
EINECS: 208-237-3
UN: 0000
Merck Index: 13,04654
Specification Test Results
Appearance Light brown powder
Em (550 nm, 0.1 M NaOH) 14700

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Amber bottle 25 g VWRC0701-25G
Amber bottle 50 g VWRC0701-50G
Amber bottle 100 g VWRC0701-100G
Expand All / Collapse All
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty