1-Fluoro-3-iodo-5-nitrobenzene ≥97%

Supplier: Thermo Scientific
Warning

ALFAH31612.03EA 0 SGD
ALFAH31612.03
1-Fluoro-3-iodo-5-nitrobenzene ≥97%
1-Fluoro-3-iodo-5-nitrobenzene
Formula: C₆H₃FINO₂
MW: 267 g/mol
Melting Pt: 77…78,5 °C
Storage Temperature: Ambient
MDL Number: MFCD00007219
CAS Number: 3819-88-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR