Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetic acid

Supplier: OT CHEMICALS
Danger



Formula: H₃CCOOH
MW: 60,05 g/mol
Boiling Pt: 118 °C (1013 hPa)
Melting Pt: 17 °C
Density: 1,05 g/cm³ (20 °C)
Flash Pt: 38,5 °C
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8,II
REACH: 01-2119475328-30
https://sg.vwr.com/stibo/web/std.lang.all/31/15/723115.jpg

Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pk VWR Catalog Number Quantity
30 kg XXXXTACE-010
Supplier Article Number:: TACE-010
Expand All / Collapse All
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty