VWRC44328
VWRC443283MEA 46.2 SGD
VWRC443283M
VWRC44328
Coomassie brilliant blue R-250 is used for post-electrophoresis staining of proteins in applications requiring a high degree of sensitivity and/or solubility.