Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zinc selenide Optical Window, 12.7mm (0.5in) dia x 2mm (0.08in) thick

Supplier: Alfa Aesar
Danger

Physical Form: Clear Aperture 85%, Surface Finish 60/40


Formula: SeZn
MW: 144,35 g/mol
Melting Pt: 1793 °C
Density: 5,4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011296
CAS Number: 1315-09-9
EINECS: 215-259-7
UN: 3283
ADR: 6.1,III
Merck Index: 12,10290

Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pk VWR Catalog Number Quantity
1 ALFA45666.KS
Supplier Article Number:: 45666.KS
Expand All / Collapse All

SPECIFICATIONS

Physical form Clear Aperture 85%, Surface Finish 60/40
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty