Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Vanadomolybdate reagent Reag. Ph. Eur. 1060100

Supplier: VWR Chemicals
Danger

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.


Boiling Pt: ~100 °C (1013 hPa)
Density: 1,2 g/cm³ (20 °C)
UN: 2031
ADR: 8,II

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Plastic bottle 200 ml VWRC87882.220
Expand All / Collapse All
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty