Would you like to log in again?
Cleaning brushes for balloon flasks
Supplier: VWR Collection
VWR®
Black nylon, wooden handle. These cleaning brushes are specially designed for cleaning balloon flasks and carboys.
  • Swivelling brush section for easy cleaning of bottoms and corners
  • For cleaning the upper inside shoulder of balloon flasks and carboys
  • Wooden handle with hanging loop
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Material Head Ø Trimming length Total length Pk VWR Catalog Number Quantity
Nylon bristle 50 mm 200 mm 550 mm 1 VWRI149-0105
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty