Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acrodisc® units with Mustang® Q and S membranes

Supplier: Pall Laboratory
Acrodisc®
Disposable chromatography units with high binding capacities and fast flow rate.

Mustang® Q, strong anion exchanger:

Provides contaminant removal such as DNA viral particles, host cell proteins or endotoxins. Ideal for isolation via capture and release of plasmid DNA, virus or target protein from a complex mixture. Offer protein polishing for negatively charged proteins. Purifies virus and oligo nucleotides.

Mustang® S, strong cation exchanger: Purifies and concentrates positively charged proteins and viral particles. Ideal for evaluation, process development and validation.

Mustang® S, strong cation exchanger:

Purifies and concentrates positively-charged proteins and viral particles. Ideal for evaluation, process development and validation.




 

ORDER

Expand All / Collapse All
Type Membrane Material Pore size Ø Sterile Pk VWR Catalog Number Quantity
Mustang® Q Mustang® Q PP 0,8 µm 25 mm - 10 PALLMSTG25Q6
Supplier Article Number:: MSTG25Q6
Mustang® S Mustang® S PP 0,8 µm 25 mm - 10 PALLMSTG25S6
Supplier Article Number:: MSTG25S6

SPECIFICATIONS

Ø (mm) 25
Pore size 0,8 µm
Inlet/outlet Female Luer lock/male Luer
Max. pressure (bar) 5,5
Flow rate 1 - 4 ml/min
Operating temperature range (°C) 21...24
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty