Would you like to log in again?
Examination gloves for sensitive skin, Micro-Touch® Hydracare®
Supplier: Ansell
Latex, non sterile, powder-free, textured finish
These latex examination gloves are formed so that they can be worn on either hand and include an inner layer that has been treated with a soothing combination of camomile and moisturisers. Ideal for daily patient examinations or dental procedures.
  • Include built-in coating to soothe sensitive skin: With camomile (cools and soothes) and moisturisers to keep skin smooth
  • The gloves are lightly perfumed to keep hands smelling fresh
  • Reliable protection against viruses, pathogens, chemicals and harsh cleaning agents, low allergenic profile (protein content less than 50 mg/g)
EN 455
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Size Colour Length Packed Pk VWR Catalog Number Quantity
XS White 240 mm 100/box 100 ANSM553331
S White 240 mm 100/box 100 ANSM553332
M White 240 mm 100/box 100 ANSM553333
L White 240 mm 100/box 100 ANSM553334
XL White 240 mm 80/box 80 ANSM553335
SPECIFICATIONS
Ambidextrous/hand-specific Ambidextrous
Disposable/Reusable Disposable
Powdered/Powder-free Powder-free
Finish Textured
EN norm EN 455
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty