Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Block heaters, analogue

Supplier: VWR Collection
These multipurpose units are ideal for incubation and activation of cultures, enzyme reactions, blood urea nitrogen determinations, immunoassays, melting or boiling points. Two separate temperature adjustment knobs provide low and high temperature control. Low range knob adjusts from above ambient to 100 °C and high range knob adjusts from 75 to 150 °C. High wattage, constant temperature dry block heaters are economical, versatile and compact. Each of the five models accepts separate, interchangeable, modular heating blocks, which accommodate various sample containers such as microtubes, centrifuge tubes, vials, microplates and PCR strips or tubes.

  • Multipurpose device for a wide range of applications
  • Temperature range from ambient +5 to 150 °C
  • Accommodate VWR® interchangeable modular heating blocks for tubes from 0,2 ml up to 50 ml capacity
  • Analogue controls

Ordering information: Heating blocks are not included, please order these separately.

Caution: * RT = Ambient
** EU-plug *** UK-plug **** CH-plug



 

ORDER

Expand All / Collapse All
Type No. of blocks Temp. range Temp. stability Heating capacity W×D×H Weight Pk VWR Catalog Number Quantity
Block heater, analogue 6 ** RT* +5...150 °C ±2,5 °C 0,41 kW 203×531×89 mm 4,5 kg 1 VWRI460-3261 Unavailable
Block heater, analogue 1 *** RT* +5...150 °C ±1,5 °C 0,11 kW 203×315×89 mm 2,6 kg 1 VWRI460-3250 Unavailable
unavailable This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +65 6505 0760.

ACCESSORIES

Expand All / Collapse All

Accessories
Description Well size For No. of holes Depth W×D×H Pk VWR Catalog Number Quantity
Single block - mm - - - 95×76×51 mm 1 VWRI460-3236
Double block - mm - - - 95×152×57 mm 1 VWRI460-3239
Single block, 64× individual PCR tubes Ø 6,4 mm 0,2 ml tubes 82 20,2 mm 95×76×51  mm 1 VWRI460-3225
Single block, 10×8-PCR tube strips Ø 6,4 mm 0,2 ml tubes 81 15,5 mm 95x76x51  mm 1 VWRU13259-256
Supplier Article Number:: 13259-256
Double block, 1×96-well PCR plate Ø 6,4 mm 0,2 ml tubes 83 15,5 mm 95×152×57  mm 1 VWRI460-3226
Single block Ø 10,9 mm 2,0 ml Corning microtubes 20 38,1 mm 95×76×51  mm 1 VWRI460-3246
Single block Ø 6,3 mm Round bottomed tubes, 6 mm Ø 30 27 mm 95×76×51  mm 1 VWRI460-3234
Single block Ø 7,9 mm 0,5 ml PCR tubes 30 27,6 mm 95×76×51  mm 1 VWRI460-3211
Single block Ø 7,9 mm 0,5 ml PCR tubes 30 27,6 mm 95×76×51  mm 1 VWRU13259-000
Supplier Article Number:: 13259-000
Single block Ø 11,1 mm 1,5 ml VWR microtubes 20 39,1 mm 95×76×51  mm 1 VWRI460-3235
Single block Ø 11,1 mm 1,5 ml VWR microtubes 20 39,1 mm 95×76×51  mm 1 VWRU13259-286
Supplier Article Number:: 13259-286
Single block Ø 11,5 mm 1,5 ml Eppendorf microtubes 20 36,9 mm 95×76×51  mm 1 VWRI460-3212
Single block Ø 11,5 mm 1,5 ml Eppendorf microtubes 20 36,9 mm 95×76×51  mm 1 VWRU13259-002
Supplier Article Number:: 13259-002
Single block Ø 11,5 mm 2,0 ml VWR/Eppendorf microtubes 20 38,1 mm 95×76×51  mm 1 VWRI460-3245
Single block Ø 11,5 mm 2,0 ml VWR/Eppendorf microtubes 20 38,1 mm 95×76×51  mm 1 VWRU12985-048
Supplier Article Number:: 12985-048
Single block Ø 17,1 mm 15 ml conical tubes 12 44,5 mm 95×76×51  mm 1 VWRI460-3221
Single block Ø 29,0 mm 50 ml conical tubes 5 47,6 mm 95×76×51  mm 1 VWRI460-3223
Single block Ø 17,1 mm 15 ml conical tubes 12 44,5 mm 95×76×51  mm 1 VWRU13259-250
Supplier Article Number:: 13259-250
Single block Ø 29,0 mm 50 ml conical tubes 5 47,6 mm 95×76×51  mm 1 VWRU13259-254
Supplier Article Number:: 13259-254
Single block Ø 8,3 mm 6 mm round bottom tubes 30 48,4 mm 95×76×51  mm 1 VWRI460-3213
Single block Ø 8,3 mm 6 mm round bottom tubes 30 48,4 mm 95×76×51  mm 1 VWRU13259-105
Supplier Article Number:: 13259-105
Single block Ø 10,7 mm 10 mm round bottom tubes 24 48,4 mm 95×76×51 mm 1 VWRI460-3214
Single block Ø 10,7 mm 10 mm round bottom tubes 24 48,4 mm 95×76×51 mm 1 VWRU13259-107
Supplier Article Number:: 13259-107
Single block Ø 13,9 mm 12/13 mm round bottom tubes 16 48,4 mm 95×76×51 mm 1 VWRI460-3215
Single block Ø 13,9 mm 12/13 mm round bottom tubes 20 48,4 mm 95×76×51 mm 1 VWRI460-3216
Single block Ø 17,5 mm 15/16 mm round bottom tubes 12 48,4 mm 95×76×51 mm 1 VWRI460-3217
Single block Ø 17,5 mm 15/16 mm round bottom tubes 12 48,4 mm 95×76×51 mm 1 VWRU13259-162
Supplier Article Number:: 13259-162
Single block Ø 19,1 mm 17/18 mm round bottom tubes 12 48,4 mm 95×76×51 mm 1 VWRI460-3243
Single block Ø 21,0 mm 20 mm round bottom tubes 8 48,4 mm 95×76×51 mm 1 VWRI460-3218
Single block Ø 26,2 mm 25 mm round bottom tubes 6 48,4 mm 95×76×51 mm 1 VWRI460-3219
Single block Ø 35,0 mm 35 mm round bottom tubes 4 47,6 mm 95×76×51 mm 1 VWRI460-3222
Single block, test tube combination 6 mm, 12/13 mm, 25 mm Ø 8,3 / Ø 13,9 / Ø 26,2 mm - 6/5/3 48,4/48,4/48,4 mm 95×76×51  mm 1 VWRI460-3220
Single block, centrifuge tube combination 1,5 ml, 15 ml, 50 ml Ø 11,1 / Ø 17,1 / Ø 29,0 mm - 4/3/2 39,1/44,5/47,6 mm 95×76×51 mm 1 VWRI460-3247
Single block, microtube combination 0,5 ml, 1,5 ml, 2,0 ml Ø 7,9 / Ø 11,1 / Ø 11,5 mm - 6/10/5 27,6/39,1/38,1 mm 95×76×51 mm 1 VWRI460-3248
Single block Ø 12,7 mm 12 mm vials 20 30,0 mm 95×76×51  mm 1 VWRI460-3279
Single block Ø 15,8 mm 15 mm vials 20 35,0 mm 95×76×51  mm 1 VWRI460-3280
Single block Ø 16,4 mm 16 mm vials 15 45,0 mm 95×76×51 mm 1 VWRI460-3287
Single block Ø 16,4 mm 16 mm vials 15 45,0 mm 95×76×51 mm 1 VWRU12985-066
Supplier Article Number:: 12985-066
Single block Ø 17,8 mm 17 mm vials 12 45,0 mm 95×76×51  mm 1 VWRI460-3281
Single block Ø 19,7 mm 19 mm vials 12 45,0 mm 95×76×51 mm 1 VWRI460-3282
Single block Ø 21,7 mm 21 mm vials 9 45,0 mm 95×76×51 mm 1 VWRI460-3283
Single block Ø 23,8 mm 23 mm vials 8 45,0 mm 95×76×51 mm 1 VWRI460-3284
Single block Ø 25,8 mm 25 mm vials 8 45,0 mm 95×76×51 mm 1 VWRI460-3285
Single block Ø 28,8 mm 28 mm vials 6 45,0 mm 95×76×51 mm 1 VWRI460-3286
Sand bath for one-block heater, 95×76×64 mm - - - - - 1 VWRI460-3227
Sand bath for two-block heater, 95×149×64 mm - - - - - 1 VWRI460-3228
Sand bath for three-block heater, 95×224×64 mm - - - - - 1 VWRI460-3229
Sand bath for four-block heater, 147×191×64 mm - - - - - 1 VWRI460-3230
Sand bath for six-block heater, 147×287×64 mm - - - - - 1 VWRI460-3231
Sand for sand bath - - - - - 450 g VWRI460-3232
Stainless steel shot for sand bath - - - - - 450 g VWRI460-3233
2-Block cover, max. tube height 89 mm - - - - 165×165×41 mm 1 VWRI460-3240
2-Block cover, max. tube height 206 mm - - - - 165×165×114 mm 1 VWRI460-3289
4-Block cover, max. tube height 89 mm - - - - 178×216×41 mm 1 VWRI460-3241
6-Block cover, max. tube height 89 mm - - - - 178×316×41 mm 1 VWRI460-3242
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping