Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Electroblotting units, BTV100

Supplier: VWR Collection
VWR®
VWR Collection electroblotting unit is designed to provide a safe and easy to operate system for the transfer of nucleic acid or proteins from a gel matrix onto a suitable transfer membrane. It can be used to blot up to four gels of 10×10 cm.

  • User-replaceable electrode consists of densely coiled platinum wire, which results in the generation of a uniform electric field, with the electrode separation calculated for optimum transfer
  • The unit has a cooling coil which is in direct contact with the buffer, and which allows coolant to be pumped through, via a chiller or from a tap, to cool the buffer
  • A stirring bar can be placed under the compression cassettes to assist in temperature control
  • Each asymmetric, colour coded compression cassette has a locking mechanism to minimise gel movement during insertion into the tank
  • The asymmetry of the cassettes prevents them being inserted into the tank in the wrong orientation, against the direction of transfer

Delivery information: Each unit is supplied with four compression cassettes and four fibre pads.



 

ORDER

Expand All / Collapse All
Description Pk VWR Catalog Number Quantity
Electroblotter, 100×100 mm 1 VWRI700-7131

SPECIFICATIONS

Model BTV100-EBK
Buffer volume (ml) 2000
Active transfer area (cm³) 71
Distance between electrodes (mm) 128
Unit W×D×H (mm) 200×225×185
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty