Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Heating circulators

Supplier: Julabo
Heating circulators with stainless steel bath tank for internal and external temperature applications. Units have powerful heating capacities for large closed or open external systems with simultaneous operation in the bath tank. Models are available with different bath sizes and operating facilities. Accessories: External PT100 sensors, bath fluids, tubing and adapters for connecting external systems. All models feature an integrated cooling coil for applications at or near ambient temperature.

  • Early warning system signals a lowering of bath fluid level
  • PID temperature control with drift compensation and electronically adjustable pump capacity
  • RS232 interface

MA models: Features include Multi-Display (LED) for actual value, up to three set points, high/low temperature warning functions and high temperature protection. It is possible to calibrate three temperatures simultaneously.
ME models: Features include Multi-Display (LED) for actual value, up to three set points, high/low temperature warning functions and high temperature protection. It is possible to calibrate three temperatures simultaneously. Units have a bright VFD comfort display for simultaneous indication of internal/external set point and actual values, and PID cascade temperature control for highest internal and external temperature stability.

Early warning system signals a lowering of bath fluid level
PID temperature control with drift compensation and electronically adjustable pump capacity
Electronically adjustable pump capacity including illuminated pump stage display
Connection for external PT100 sensor for measurement and control
Integrated controller with 10 program steps
RS232 interface

HE, SE models: Features as ME, and in addition have optional electronic module with analogue connections for temperature recorder, set point programmer.
ICC (Intelligent Cascade Control), self-optimising temperature control, for optimum results
TCF (Temperature Control Features) for band limit, limit setting, co-speed factor and control dynamics
Particularly powerful pressure and suction pump to 1,1 bar, electronically adjustable

HL, SL models: Features as HE/SE, and in addition have LCD dialogue display, backlit with interactive menu and connections for optional solenoid valves or HST Booster heater with an additional heating capacity of 4 kW.
RS232/RS485 interface for comprehensive communication and networking possibilities
Integrated programmer for 6×60 program steps

Max. flow rate / max. pressure:
ME: 11 - 16 l/min, 450 mbar
HE, SE, HL, SL: 22 - 26 l/min, 700 mbar pressure, 400 mbar suction

Certifications: Classification (according to DIN 12876-1: 3): MA, ME, HE, SE, HL, SL

Ordering information: All units with an RS232 interface can use the free EasyTemp software for control via a PC; the software can be downloaded free of charge. Please contact your local VWR office for details.




 

ORDER

Expand All / Collapse All
Type Temp. range Temp. stability Heating capacity Capacity Bath W×D×H W×D×H Pk VWR Catalog Number Quantity
ME-4 20...200 °C ±0,01 °C 2 kW 4,5 l 130×150×150 mm 210×420×380 mm 1 JULA9162504
Supplier Article Number:: 9162504
ME-6 20...200 °C ±0,01 °C 2 kW 6 l 130×150×200 mm 210×430×420 mm 1 JULA9162506
Supplier Article Number:: 9162506
ME-12 20...200 °C ±0,01 °C 2 kW 12 l 220×150×200 mm 300×430×450 mm 1 JULA9162512
Supplier Article Number:: 9162512
ME-26 20...200 °C ±0,01 °C 2 kW 26 l 220×300×200 mm 360×610×450 mm 1 JULA9162526
Supplier Article Number:: 9162526
HE-4 20...250 °C ±0,01 °C 2 kW 4,5 l 130×150×150 mm 210×420×400 mm 1 JULA9212504
Supplier Article Number:: 9212504
SE-6 20...300 °C ±0,01 °C 3 kW 6 l 130×150×200 mm 210×430×440 mm 1 JULA9252506
Supplier Article Number:: 9252506
SE-12 20...300 °C ±0,01 °C 3 kW 12 l 220×150×200 mm 300×430×470 mm 1 JULA9252512
Supplier Article Number:: 9252512
SE-26 20...300 °C ±0,01 °C 3 kW 26 l 220×300×200 mm 360×610×470 mm 1 JULA9252526
Supplier Article Number:: 9252526
HL-4 20...250 °C ±0,01 °C 2 kW 4,5 l 130×150×150 mm 210×420×400 mm 1 JULA9312504
Supplier Article Number:: 9312504
SL-6 20...300 °C ±0,01 °C 3 kW 6 l 130×150×200 mm 210×430×440 mm 1 JULA9352506
Supplier Article Number:: 9352506
SL-12 20...300 °C ±0,01 °C 3 kW 12 l 220×150×200 mm 300×430×470 mm 1 JULA9352512
Supplier Article Number:: 9352512
SL-26 20...300 °C ±0,01 °C 3 kW 26 l 220×150×200 mm 360×610×470 mm 1 JULA9352526
Supplier Article Number:: 9352526
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping