Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Biological safety cabinets, Class II, Herasafe™, KS series

Supplier: Thermo Scientific
HERAsafe®
These biological safety cabinets offer high levels of safety for users and product. They are ideal for use in fields such as biotechnology, microbiology, research, laboratories, pharmacies and hospitals. They are compliant with safety levels 1 - 3 as required in microbiology, biotechnology and gene technology applications.

  • Aerosol-tight electric front window provides ease of sealing for fumigation and a clean sterile environment when the window is closed
  • Sash opening: 254 mm
  • Main and exhaust HEPA filter 99.999% efficiency for 0,3 µm particles (type H14 according to EN 1822) pre-stage filter 85% efficiency for 5 µm particles

Their ergonomic design allows unobstructed, all-round visibility with frameless front window and transparent glass side windows. The front opening is over 250 mm to enable convenient handling. Corners on the inside are rounded and windows can be folded up completely for easy cleaning. Air supply and exhaust is controlled automatically by the ventilation system. The display on the back wall of the workspace is placed at eye level, so that all parameters and operating functions can be monitored comfortably. Optional connections for gas or other media are provided on the side or rear walls. These cabinets are equipped with ergonomically designed arm rests.

With advanced internal airflow and motor technology, HERAsafe® KS cabinets offer optimal energy efficiency, operation and serviceability. At full speed the motors consume 60% less energy than standard cabinets. To further conserve energy, the intelligent speed control automatically reduces blower speed to 30% when the window is closed. This technology extends HEPA filter life while ensuring a sterile working environment and saves energy to minimise operating costs.

Certifications: Certified according to EN NF 12469.

Ordering information: A wide selection of support stands and alternative work surfaces, which also provide the option of using a balance, UV lamps, sockets, gas connections and other components are available to adapt the cabinets to your needs.

Delivery information: Each cabinet is supplied with two electrical sockets, one on each side wall and 4 ports for connection of utility taps.
Delivery, installation and commissioning prices available on request. Please contact VWR for details.


 

ORDER

Expand All / Collapse All
Description Pk VWR Catalog Number Quantity
Class II cabinet, HERAsafeTM KS12 1 HERA51022482
Supplier Article Number:: 51022482
Class II cabinet, HERAsafeTM KS15 1 HERA51022483
Supplier Article Number:: 51022483
Class II cabinet, HERAsafeTM KS18 1 HERA51022484
Supplier Article Number:: 51022484
Class II cabinet, HERAsafeTM KS9 1 HERA51022481
Supplier Article Number:: 51022481

SPECIFICATIONS

Model KS12 KS15 KS9 KS18
Exhaust air volume (m³/h) 500 625 380 750
Lighting system (lux) >1000
Noise level (dB(A)) 57 58 57 58
Power rating (kW) 0,4 0,8 0,4 0,8
Ext. W×D×H (mm) 1300×800×1645 1600×800×1645 1000×800×1645 1900×800×1645
Int. W×D×H (mm) 1200×627×780 1500×627×780 900×627×780 1800×627×780
Weight (kg) 240 290 190 340
Filters HEPA H14 EN 1822, 99,999% at 0,3 μm particle size
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty