Would you like to log in again?
Multi-channel pipettes, electronic, variable volume
Supplier: VWR Collection
VWR®
These lightweight, ergonomically designed micropipettes prevent hands from becoming tired during repetitive operations. They are equipped with a microprocessor controlled stepper motor for dispensing precise volumes with the help of a piston and an optical coupler that monitors the position of the piston to guarantee that the movement is repeated with precision every time.
  • Wide LCD screen and convenient interface
  • Memory is capable of storing up to nine configurations of functional parameters
  • Permanent self-test programme ensures the precision of the pipetted volumes and battery dependence
  • Auto-calibration function
Ordering information: Supplied with one lithium-ion battery, one clip for fixing to a shelf, and one adapter for recharging the battery in the micropipette.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
8-Channel
Capacity Division Accuracy Imprecision With Pk VWR Catalog Number Quantity
2 - 20 µl 0,1 µl ±5,0 - ±1,0 % <2,0 - <0,3 % EU-plug 1 VWRI613-0410 Unavailable
12-Channel
Capacity Division Accuracy Imprecision With Pk VWR Catalog Number Quantity
10 - 200 µl 1,0 µl ±2,0 - ±0,6 % <1,0 - <0,15 % EU-plug 1 VWRI613-0420 Unavailable
unavailable This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +65 6505 0760.
ACCESSORIES
Expand All / Collapse All
Accessories
Description Pk VWR Catalog Number Quantity
Acrylic stand for 3 electronic single channel or multi channel pipettes 1 VWRI613-0428
Carousel for 6 electronic single channel or multi channel pipettes 1 VWRI613-0429
Lithium-ion battery 1 VWRI613-0424
Shelf clip for 1 electronic single channel or multi channel pipette 1 VWRI613-0427
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty