Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cooling/heating microtube dry blocks, Eppendorf ThermoStat™ plus

Supplier: Eppendorf
User friendly ThermoStat™ plus gives accurate temperature control for all commonly used laboratory tubes. Applications include enzyme reactions, transformation, denaturation of DNA, RNA and proteins, storage of enzymes or nucleic acids at defined temperatures, cell lysis, in vitro translation, ligation and hybridisation.

  • Peltier technology gives precise temperature control with active cooling and heating
  • Individual programming of up to four separate time phases and temperature ramps
  • Run time for linear transition between two temperature levels can be individually programmed
  • Timer function: 1 min to 99,59 h or continuous operation
  • RS232 interface for remote control and documentation

IP protection class according to DIN EN 61010: IP 20

Ordering information: Supplied without thermoblocks, please order required thermoblocks separately.



 

ORDER

Expand All / Collapse All
Description W×D×H Weight Pk VWR Catalog Number Quantity
ThermoStat plus, EU-plug 220×250×125 mm 5 kg 1 EPPE5352000.010
Supplier Article Number:: 5352000.010
ThermoStat™ plus, UK-plug 220×250×125 mm 5 kg 1 EPPE5352000.037
Supplier Article Number:: 5352000.037

SPECIFICATIONS

Temperature setting (°C) From -5 to +99
Incubating accuracy (°C) ±1.0 at 0 °C
±0.5 at 37 °C
±1.0 at 90 °C
±1,0 at 0 °C
±0,5 at 37 °C
±1,0 at 90 °C
Heating speed (°C/min) 5
Cooling speed (°C/min) 6 between 99 and 25 °C
1.5 between 25 and -5 °C
6 between 99 and 25 °C
1,5 between 25 and -5 °C
Temperature control range (°C) * From 30 below ambient to 99 °C
Motor input  (W) 75
Peltier input (W) 75
W×D×H (mm) 220×250×125 mm 220×250×125
Weight (kg) 5 kg

ACCESSORIES

Expand All / Collapse All

Accessories
Description For Max. revolutions Pk VWR Catalog Number Quantity
Adapter plate for 96×0,2 ml PCR tubes and PCR plates 70 °C maximum 1400 min-1 1 EPPE5363007.009
Supplier Article Number:: 5363007.009
Combibox for all tube types, interior 135×90×60 mm 95 °C maximum - 1 EPPE5352100.007
Supplier Article Number:: 5352100.007
Interchangeable thermoblock for 24×0,5 ml microcentrifuge tubes 99 °C maximum 1500 min-1 1 EPPE5361000.015
Supplier Article Number:: 5361000.015
Interchangeable thermoblock for 24×1,5 ml microcentrifuge tubes 99 °C maximum 1400 min-1 1 EPPE5360000.011
Supplier Article Number:: 5360000.011
Interchangeable thermoblock for 24×2,0 ml microcentrifuge tubes 99 °C maximum 1400 min-1 1 EPPE5362000.019 Unavailable
Supplier Article Number:: 5362000.019
Interchangeable thermoblock for 4×50 ml conical tubes 99 °C maximum 750 min-1 1 EPPE5365000.010
Supplier Article Number:: 5365000.010
Interchangeable thermoblock for 4 Eppendorf DualChip slides, with clamping plate 70 °C maximum 1400 min-1 1 EPPE5368000.100
Supplier Article Number:: 5368000.100
Interchangeable thermoblock for 4 slides for hybridisation experiments 99 °C maximum 1400 min-1 1 EPPE5368000.010
Supplier Article Number:: 5368000.010
Interchangeable thermoblock for microtitre and deep well plates, PCR tubes and plates, with lid 99 °C maximum 1400 min-1 1 EPPE5363000.012
Supplier Article Number:: 5363000.012
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty