Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Leishman Stain

Supplier: Alfa Aesar
CAS No.: 12627-53-1
Odour: Odourless
MDL No.: MFCD00131498



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pk Pack type VWR Catalog Number Quantity
25 g Glass bottle for solids ALFAA11644.14
Supplier Article Number:: A11644.14
100 g Glass bottle for solids ALFAA11644.22
Supplier Article Number:: A11644.22

SPECIFICATIONS

CAS No. 12627-53-1
Odor/Scent Odourless
MDL No. MFCD00131498
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty