Powered air purifying respirator (PAPR) system, Breathe Easy™
Supplier: 3M
Breathe Easy™
MMMA520-15-00EA 2250 SGD
MMMA520-15-00 MMMA520-17-00
Powered air purifying respirator (PAPR) system, Breathe Easy™
Respirators Respiratory Systems
PAPR system provides comfortable and efficient powered air protection. Ambient air is drawn through the filter cartridge, supplying filtered air through the breathing tube to the headpiece.
  • Headpieces are constructed of Tychem® QC for protection from chemicals; they are available in hood, sealed-seam hood, or head cover styles
  • Loose-fitting hood and sealed-seem hood accommodate facial hair and glasses
  • Lightweight head cover is available in white or blue
  • Sealed-seam hood assembly 111-0647 features a built-in collar and cap suspension
  • Sealed-seam hood assembly 111-0651 features a built-in collar and hardha
  • Assemblies include motor/blower, battery pack, belt, and flowmeter
  • Assembly 111-0627 is 'intrinsically safe' (i.e., incapable of creating a spark or explosion)
  • Assemblies use interchangeable cartridges/filters that protect against gases, vapors, or particulates
Certifications: NIOSH approved.
Ordering information: PAPR assembly, headpiece, smart charger, filters, and breathing tube are ordered separately.
Caution: All 3M™ products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.