Sodium dihydrogen phosphate monohydrate, Electran for molecular biology
Synonyms:
Sodium Phosphate Monobasic hydrate
, Sodium dihydrogen orthophosphate monohydrate
, Sodium phosphate monobasic monohydrate
VWRC444435XEA
127.64
SGD
VWRC444435X
Sodium dihydrogen phosphate monohydrate, Electran for molecular biology
Sodium dihydrogen phosphate monohydrate
Formula:
NaH₂PO₄·H₂O MW: 137,99 g/mol Melting Pt: 100 °C Density: 2,04 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149208 CAS Number: 10049-21-5 EINECS: 231-449-2 Merck Index: 12,08806 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...