Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Barium chloride dihydrate

SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
 
 
SearchResultCount:"10"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VWR Chemicals
Description:   Barium chloride dihydrate ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier:  VWR Chemicals
Description:   BARIUM CHLORIDE DIHYDRATE ACS GRADE 1 * 500 g
Catalog Number: (VWRC21709.364)
Supplier:  VWR Chemicals
Description:   Barium chloride dihydrate TECHNICAL
UOM:  1 * 5 kg
Supplier:  VWR Chemicals
Description:   Barium chloride dihydrate ≥99%, GPR RECTAPUR®
Supplier:  Avantor Performance Materials
Description:   Crystals. Lot analysis on label.
Catalog Number: (BAKR0974-04)
Supplier:  Avantor Performance Materials
Supplier Article Number:: 0974-04
Description:   Crystals. For Parr turbidimeter. Lot analysis on label.
UOM:  1 * 125 g
Supplier:  Alfa Aesar
Description:   Barium chloride dihydrate ≥99%
Supplier:  Alfa Aesar
Description:   Barium chloride dihydrate ≥99.997%, Puratronic®
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +65 6505 0760.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +65 6505 0760.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR at sales@sg.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +65 6505 0760.
1 - 8  of 10
  1  2  Next

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping