Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "Calcium nitrate tetrahydrate"

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"7"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VWR Chemicals
Description:   Calcium nitrate tetrahydrate AnalaR NORMAPUR® analytical reagent
UOM:  1 * 1 kg
Supplier:  VWR Chemicals
Description:   Calcium nitrate tetrahydrate GPR RECTAPUR®
UOM:  1 * 1 kg
Supplier:  VWR Chemicals
Description:   CALCIUM NITRATE TETRAHYDRATE GRAN 1 * 500 g
Catalog Number: (VWRC22384.367)
Supplier:  VWR Chemicals
Description:   Calcium nitrate tetrahydrate TECHNICAL
UOM:  1 * 5 kg
Supplier:  Alfa Aesar
Description:   Calcium nitrate tetrahydrate Puratronic®
Supplier:  Alfa Aesar
Description:   Calcium nitrate tetrahydrate
Supplier:  Alfa Aesar
Description:   Calcium nitrate tetrahydrate
UOM:  1 * 500 g
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +65 6505 0760.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +65 6505 0760.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR at sales@sg.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +65 6505 0760.
1 - 7  of 7