Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lanthanum(III) nitrate hydrate, (H₂O 22-25%) ≥99%

Supplier: Alfa Aesar
Danger



Formula: H₂LaN₃O₁₀
MW: 342,932 g/mol
Boiling Pt: 126 °C (1013 hPa)
Melting Pt: 40 °C
Density: 2,347 g/cm³ (20 °C)
Storage Temperature: Ambient
CAS Number: 100587-94-8
EINECS: 233-238-0
UN: 1477
ADR: 5.1,III
Merck Index: 14,05363

 

ORDER

Expand All / Collapse All
Pk VWR Catalog Number Quantity
100 g ALFAA14890.22
Supplier Article Number:: A14890.22
500 g ALFAA14890.36
Supplier Article Number:: A14890.36
Expand All / Collapse All

SPECIFICATIONS

Melting point 40°
Formula weight 324.92(anhy)
Molecular formula LaN3O9.xH2O
MDL No. MFCD00011067
UN No. 1477
Merck reference 14,5363
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping