Pelargonaldehyde ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
1-Nonanal
, Nonylaldehyde
, Nonaldehyde
, Nonanal
ALFAA15908.36EA
539
SGD
ALFAA15908.36
ALFAA15908.22
Pelargonaldehyde ≥97%
Pelargonaldehyde
Formula:
C₉H₁₈O MW: 142,24 g/mol Boiling Pt: 79…81 °C (12 mmHg) Melting Pt: –40 °C Density: 0,825 Flash Pt: 63 °C (145 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007030 CAS Number: 124-19-6 EINECS: 204-688-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...