Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetonitrile, HPLC Grade, 99.7+% min

Supplier: Alfa Aesar

Purity: 99.7+% min
CAS No.: 75-05-8
Molecular Formula: CH3CN
Formula Weight: 41.05
Flash Point: 5°(41°F)
Physical Form: Liquid
Density: 0.786
Refractive Index: 1.3440
Odour: Pungent
Melting Point: -48° to -45°
Boiling Point: 80-82°
Beilstein Reference: 741857
Merck Reference: 14,70
UN No.: 1648
MDL No.: MFCD00001878

(Cyanomethane Ethanenitrile Methyl cyanide Ethanoic acid nitrile)
R: 11-20/21/22-36
S: 16-36/37
H: H225 H302+H312+H332 H319
P: P210 P280 P305+P351+P338 P309+P310
Danger
 
  Formula: H₃CCN
MW: 41,05 g/mol
Boiling Pt: 81.6 °C (1013 hPa)
Melting Pt: -45.7 °C
Density: 0,782 g/cm³ (20 °C)
Flash Pt: 2 °C (closed cup)
MDL Number: MFCD00001878
CAS Number: 75-05-8
ADR: 3,II
REACH: 01-2119471307-38
https://sg.vwr.com/stibo/web/std.lang.all/89/10/718910.jpg
Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle 1 l ALFA22927.K2
Expand All / Collapse All

SPECIFICATIONS

Density 0.786
Melting Point -48° to -45°
Formula Weight 41.05
CAS No. 75-05-8
Purity 99.7+% min
Refractive Index 1.3440
Flash Point 5°(41°F)
Physical Form Liquid
Odor/Scent Pungent
Boiling Point 80-82°
Molecular Formula CH3CN
MDL No. MFCD00001878
UN No. 1648
Merck Reference 14,70
Beilstein Reference 741857
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty