Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-5-(trifluoromethyl)benzenesulphonyl chloride ≥97%

Supplier: Alfa Aesar
Danger
Synonyms: 2-Chloro-5-(trifluoromethyl)benzene-1-sulphonyl chloride



Formula: C₇H₃Cl₂F₃O₂S
MW: 279,07 g/mol
Boiling Pt: 117…119 °C (1,0 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00052310
CAS Number: 54090-08-3
UN: 3265
ADR: 8,II

 

ORDER

Expand All / Collapse All
Pk VWR Catalog Number Quantity
1 g ALFAB20476.03
Supplier Article Number:: B20476.03
5 g ALFAB20476.06
Supplier Article Number:: B20476.06
Expand All / Collapse All

SPECIFICATIONS

Formula weight 279.07
CAS No. 54090-08-3
Purity 97%
Boiling point 117-119°/1.0mm
Molecular formula C7H3Cl2F3O2S
MDL No. MFCD00052310
UN No. 3265
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty