Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-5-iodo-6-methylpyridine ≥95%

Supplier: Alfa Aesar
Danger



Formula: C₆H₅ClIN
MW: 253,47 g/mol
Melting Pt: 30…33 °C
Storage Temperature: Ambient
MDL Number: MFCD09037461
CAS Number: 249291-79-0

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 1 g ALFAH27656.03
Supplier Article Number:: H27656.03
Glass bottle for solids 250 mg ALFAH27656.MD
Supplier Article Number:: H27656.MD
Expand All / Collapse All

SPECIFICATIONS

Melting point 30-33°
Formula weight 253.47
CAS No. 249291-79-0
Purity 95%
Molecular formula C6H5ClIN
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty