Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-6-(trifluoromethyl)benzoic acid, 97%

Supplier: Alfa Aesar

Purity: 97%
CAS No.: 2376-00-3
Molecular Formula: C8H4ClF3O2
Formula Weight: 224.57
Melting Point: 123-125°
MDL No.: MFCD06208254

(6-Chloro-α,α,α-trifluoro-o-toluic acid)
 
  Formula: C₈H₄ClF₃O₂
MW: 224,57 g/mol
CAS Number: 2376-00-3
Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 5 g ALFAH30490.06
Glass bottle for solids 1 g ALFAH30490.03
Expand All / Collapse All

SPECIFICATIONS

Melting Point 123-125°
Formula Weight 224.57
CAS No. 2376-00-3
Purity 97%
Molecular Formula C8H4ClF3O2
MDL No. MFCD06208254
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty