Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Chloro-2-fluoro-3-iodopyridine ≥95%

Supplier: Alfa Aesar
Warning



Formula: C₅H₂ClFIN
MW: 257,43 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12922962
CAS Number: 1187732-65-5

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 250 mg ALFAH32895.MD
Supplier Article Number:: H32895.MD
Expand All / Collapse All

SPECIFICATIONS

Formula weight 257.43
CAS No. 1187732-65-5
Purity 95%
Molecular formula C5H2ClFIN
MDL No. MFCD12922962
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty