Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl-2-ethylacetoacetate ≥97%

Supplier: Alfa Aesar
Warning



Formula: C₇H₁₂O₃
MW: 144,17 g/mol
Boiling Pt: 189-191 °C
Melting Pt: -28 °C
Density: 1,010
Storage Temperature: Ambient
MDL Number: MFCD01632136
CAS Number: 51756-08-2
EINECS: 257-381-3

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 100 g ALFAB21368.22
Supplier Article Number:: B21368.22
Glass bottle for solids 25 g ALFAB21368.14
Supplier Article Number:: B21368.14
Expand All / Collapse All

SPECIFICATIONS

Formula weight 144.17
CAS No. 51756-08-2
Purity 97%
Refractive index 1.423
Boiling point 189-191°
Molecular formula C7H12O3
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping