Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Fluoro-5-(trifluoromethyl)benzenesulphonyl chloride ≥97%

Supplier: Alfa Aesar
Synonyms: 2-Fluoro-5-(trifluoromethyl)benzene-1-sulphonyl chloride

Purity: 97%
CAS No.: 1744-43-0
Molecular Formula: C7H3ClF4O2S
Formula Weight: 262.61
Refractive Index: 1.4780


Formula: C₇H₃ClF₄O₂S
MW: 262,61 g/mol
CAS Number: 1744-43-0
UN: 3265
ADR: 8,II


 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 250 mg ALFAH26687.MD
Supplier Article Number:: H26687.MD
Glass bottle for solids 1 g ALFAH26687.03
Supplier Article Number:: H26687.03
Expand All / Collapse All

SPECIFICATIONS

Formula weight 262.61
CAS No. 1744-43-0
Purity 97%
Refractive index 1.4780
Molecular formula C7H3ClF4O2S
MDL No. MFCD01090999
UN No. 3265
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty