Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Fluoro-5-(trifluoromethyl)pyridine ≥97%

Supplier: Alfa Aesar
Warning



Formula: C₆H₃F₄N
MW: 165,09 g/mol
Boiling Pt: 118-122 °C
Density: 1,371
Flash Pt: 35 °C (95 °F)
Storage Temperature: Ambient
MDL Number: MFCD07437945
CAS Number: 69045-82-5
EINECS: 400-290-2
ADR: 3,III

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 10 g ALFAH32574.09
Supplier Article Number:: H32574.09
Glass bottle for solids 1 g ALFAH32574.03
Supplier Article Number:: H32574.03
Expand All / Collapse All

SPECIFICATIONS

Formula weight 165.09
CAS No. 69045-82-5
Purity 97%
Refractive index 1.4020
Flash point 35°(95°F)
Boiling point 118-122°
Molecular formula C6H3F4N
MDL No. MFCD07437945
UN No. 1993
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping