Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Bromo-5-(trifluoromethyl)benzaldehyde ≥97%

Supplier: Alfa Aesar
Warning



Formula: C₈H₄BrF₃O
MW: 253,02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08059506
CAS Number: 477535-41-4

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 5 g ALFAH26544.06
Supplier Article Number:: H26544.06
Glass bottle for solids 1 g ALFAH26544.03
Supplier Article Number:: H26544.03
Expand All / Collapse All

SPECIFICATIONS

Formula weight 269.01
CAS No. 886498-07-3
Purity 97%
Refractive index 1.4990
Molecular formula C8H4BrF3O2
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty