3-Chloro-2-methylbenzenesulphonyl chloride ≥98%
|
Danger
|
Synonyms:
3-Chloro-2-methylbenzene-1-sulphonyl chloride
ALFAA13322.06EA
333
SGD
ALFAA13322.06
ALFAA13322.03
3-Chloro-2-methylbenzenesulphonyl chloride ≥98%
3-Chloro-2-methylbenzenesulphonyl chloride
Formula:
C₇H₆Cl₂O₂S MW: 225,09 g/mol Melting Pt: 69…73 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052198 CAS Number: 80563-86-6 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...