Heptafluorobutyramide ≥97%

Supplier: Thermo Scientific
Warning

ALFAB21466.09EA 0 SGD
ALFAB21466.09 ALFAB21466.18
Heptafluorobutyramide ≥97%
Heptafluorobutyramide
Formula: C₄H₂F₇NO
MW: 213,05 g/mol
Melting Pt: 102…106 °C
Storage Temperature: Ambient
MDL Number: MFCD00039773
CAS Number: 662-50-0
EINECS: 211-553-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR