Pour out connections for flasks, DURAN®
Supplier: DWK Life Sciences
DURAN®
SCOT1016425EA 23.3 SGD
SCOT1016425
Pour out connections for flasks, DURAN®
Adapters Adapters, Conical Ground Joints
DURAN®, borosilicate glass 3.3, clear, with KECK clip for conical joints.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes