Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,3,4,4,5,5,5-Heptafluoro-2-pentanone ≥97%

Supplier: Alfa Aesar

Purity: 97%
CAS No.: 355-17-9
Molecular Formula: C5H3F7O
Formula Weight: 212.07
Boiling Point: 63-64°
UN No.: 1224
MDL No.: MFCD00042079

Formula: C₅H₃F₇O
MW: 212,07 g/mol
Boiling Pt: 61 - 62 °C (1013 hPa)
Density: 1,424 g/cm³ (20 °C)
MDL Number: MFCD00042079
CAS Number: 355-17-9
EINECS: 206-577-7


Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 1 g ALFAL16804.03
Supplier Article Number:: L16804.03
Glass bottle for solids 5 g ALFAL16804.06
Supplier Article Number:: L16804.06
Expand All / Collapse All

SPECIFICATIONS

Formula Weight 212.07
CAS No. 355-17-9
Purity 97%
Boiling Point 63-64°
Molecular Formula C5H3F7O
MDL No. MFCD00042079
UN No. 1224
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty