Would you like to log in again?
Culture flasks, Corning® Synthemax™ surface
Supplier: Corning®
Corning®
The Corning® Synthemax™ surface is designed to mimic a cells’ natural environment with an extracellular matrix derived cell adhesion promoting peptide, offering a synthetic alternative to biological coatings and/or feeder cell layers used in traditional stem cell culture. The proprietary surface coating technology ensures lot-to-lot consistency, facilitating reproducible results.
  • Gamma sterilised (SAL 10-6)
  • Quality tested for lot-to-lot consistency and traceability
  • Stored at room temperature for 2 years
  • Ready to use
The Corning® Synthemax™-R peptide acrylate surface creates a uniform active surface for stem cell attachment, growth and differentiation, especially in chemically defined media. The Corning® Synthemax™-T surface supports the expansion of stem cells in their undifferentiated state and enables directed differentiation into specialised cell types. Corning® Synthemax™-T surface allows for the scalable expansion of hESC cultures for more than 20 serial passages in defined xeno-free medium.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Synthemax™-R surface
Capacity Cap Growth area Neck Recommended working volume Packed Pk VWR Catalog Number Quantity
60 Vented 75 canted 15-22,5 2/case 2 CORN3983
Supplier Article Number:: 3983
Synthemax™-T surface
Capacity Cap Growth area Neck Recommended working volume Packed Pk VWR Catalog Number Quantity
370 Vented 225 Angled 45-67,5 mL 2/case 2 CORN3976
Supplier Article Number:: 3976
60 Vented 75 canted 15-22,5 2/case 2 CORN3972
Supplier Article Number:: 3972
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty