Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Biological safety workbenches, Class II, series KSP, HERAsafe®

Supplier: Thermo Scientific
HERAsafe®
These microbiological safety workbenches offer superior personnel and product protection with best-in-class airflow and filter technology; exceptionally comfortable ergonomic design that prevents user fatigue and promotes safe working habits; and maximised energy efficiency for environmental protection and significant cost savings. They are ideal for pharmacies preparing cytotoxic drugs and in pharmaceutical control laboratories.

  • Aerosol-tight electric front window provides ease of sealing for fumigation and a clean sterile environment when the window is closed
  • 100 000 times better air filtration
  • HEPA filters guarantee maximum filtration efficiency of 99,995% at the most penetrating particle size (MPPS)
  • SmartFlow digital technology maintains constant airflow during normal filter loading or temporary airflow obstruction due to foreign objects
  • Independent alarm system immediately alerts to any airflow obstruction
  • Display parameters include downflow air velocity, cabinet malfunction, time of day, operating time of optional UV-C lights, total operating hours
  • Scratch resistant stainless-steel work surface with indentations that trap spilt liquids

KSP models offer all the benefits of the HERAsafe® KS, with added protection for hazardous applications. An additional segmented H14 HEPA filter system is used directly below the work area of the HERAsafe® KSP, protecting inner air ducts, plenums and fans from contamination and providing a cumulative filtration efficiency of 99.99999975% to the downflow and exhaust airflow.

Their ergonomic design allows unobstructed, all-round visibility with frameless motorised front window and transparent glass side windows. The front opening is over 200 mm to enable convenient handling. Corners are rounded onthe inside and windows can be folded up completely for easy cleaning. Air supply and exhaust is controlled automatically by the ventilation system. The display on the back wall of the workspace is placed at eye level, so that all parameters and operating functions can be monitored comfortably. Optional connections for gas or other media are provided on the side or rear walls. These cabinets are equipped with ergonomically designed arm rests.

Certifications: Certified according to EN12469 by TÜV Nord. The HERAsafe® KSP biological safety cabinets meet the latest European and worldwide standards for applications with stringent safety requirements.

Ordering information: A wide selection of support stands and alternative work surfaces, which also provide the option of using a balance, UV lamps, sockets, gas connections and other components are available to adapt the cabinets to your needs.

Delivery information: Delivery, installation and commissioning prices available on request. Please contact the VWR Technical Office.

Packaging: Each cabinet is supplied with two electrical sockets, one in each side wall, floor stand and 4 ports for connecting utility taps.


 

ORDER

Expand All / Collapse All
Description Pk VWR Catalog Number Quantity
Class II cabinet, with triple filter system, HERAsafe® KSP9 1 HERA51023606
Supplier Article Number:: 51023606
Class II cabinet with triple filter system, HERAsafe® KSP12 1 HERA51023607
Supplier Article Number:: 51023607
Class II cabinet with triple filter system, HERAsafe® KSP15 1 HERA51023608
Supplier Article Number:: 51023608
Class II cabinet with triple filter system, HERAsafe® KSP18 1 HERA51023609
Supplier Article Number:: 51023609

SPECIFICATIONS

Model KSP12 KSP9 KSP 15 KSP 18
Exhaust air volume (m³/h) 500 375 628 754
Lighting system (lux) >850 >800 >1250 >1300
Noise level (dB(A)) 56 58
Power rating (kW) 0,4 0,7
Ext. W×D×H (mm) 1300×870×1580 1000×870×1580 1600×870×1580 1900×870×1580
Int. W×D×H (mm) 1200×465×780 900×465×780 1500×465×780 1800×465×780
Weight (kg) 240 200 280 330
Filters HEPA H14 EN 1822, 99,995% at the most penetrating particle size

ACCESSORIES

Expand All / Collapse All

Accessories
Description Pk For VWR Catalog Number Quantity
Direct duct exhaust transition for 1,2 and 1,5 m cabinet, connection diameter 200 m 1 - HERA50073771
Supplier Article Number:: 50073771
Direct duct exhaust transition for 1,8 m cabinet, connection diameter 200 mm 1 - HERA50078790
Supplier Article Number:: 50078790
Thimble duct exhaust transition for 1,2 and 1,5 m cabinet, connection diameter 200 mm 1 - HERA50073770
Supplier Article Number:: 50073770
Thimble duct exhaust transition for 1,8 m cabinet, connection diameter 200 mm 1 - HERA50078795
Supplier Article Number:: 50078795
Thimble duct exhaust transition for 0,9 m cabinet. Connection diameter 200 mm 1 - HERA50078796
Supplier Article Number:: 50078796
Portable UV irradiator 1 Mobile UV light on stand HERA50048696
Supplier Article Number:: 50048696
UV socket for mobile UV light option 1 - HERA51900643
Supplier Article Number:: 51900643
Mobile UV light, hanging from rear wall 1 - HERA50073962
Supplier Article Number:: 50073962
UV socket for mobile UV light option, for retro-fit in exisiting cabinet 1 - HERA50109996
Supplier Article Number:: 50109996
HEPA filter for 0,9 m cabinet, field installed 1 - HERA50078857
Supplier Article Number:: 50078857
HEPA filter for 1,8 m cabinet, field installed 1 - HERA50078856
Supplier Article Number:: 50078856
Carbon filter for 0,9 m cabinet, field installed 1 - HERA50078783
Supplier Article Number:: 50078783
Carbon filter for 1,8 m cabinet, field installed 1 - HERA50078784
Supplier Article Number:: 50078784
Activated carbon filter, fit under 0,9 m cabinet worksurface, KSP models only 1 - HERA51900781
Supplier Article Number:: 51900781
Powder coated rear wall instead of stainless steel 1 - HERA51900648
Supplier Article Number:: 51900648
Left side wall, stainless steel plate instead of glass 1 - HERA51900644
Supplier Article Number:: 51900644
Right side wall, stainless steel plate instead of glass 1 - HERA51900645
Supplier Article Number:: 51900645
Left side wall polycarbonate instead of glass 1 - HERA51900646
Supplier Article Number:: 51900646
Right side wall polycarbonate instead of glass 1 - HERA51900647
Supplier Article Number:: 51900647
Vibration damped work surface, stainless steel, with granite plate, for weighing equipment and microscopy 1 Microbiological Safety Cabinets, HERAsafe® HERA50073757
Supplier Article Number:: 50073757
Water tap for rear wall, green 1 - HERA50068233
Supplier Article Number:: 50068233
Vacuum tap for rear wall, grey 1 - HERA50068232
Supplier Article Number:: 50068232
Non-combustible gas tap for rear wall, green 1 - HERA50068234
Supplier Article Number:: 50068234
Combustible gas tap for rear wall, yellow 1 - HERA50068235
Supplier Article Number:: 50068235
Vibration damped work surface 600 mm width. Constructed in stainless steel with granite plate. 1 - HERA50076162
Supplier Article Number:: 50076162
Grid pre-filter for 0,9 and 1,2 m 5 - HERA50076846
Supplier Article Number:: 50076846
Additional exhaust carbon filter for all models, factory installed 1 - HERA51900658
Supplier Article Number:: 51900658
Additional exhaust HEPA filter for 1,2 and 1,5 m cabinet, field installed 1 - HERA50073772
Supplier Article Number:: 50073772
Additional exhaust Carbon filter for 1,2 and 1,5 m cabinet, field installed 1 - HERA50073773
Supplier Article Number:: 50073773
Area module working surface, AMW3, 300 mm wide, weighing area 260×390 mm 1 - HERA50047489
Supplier Article Number:: 50047489
Area module working surface, AMW6, 600 mm wide, weighing area 560×390 mm 1 - HERA50047502
Supplier Article Number:: 50047502
Worksurface module, One-piece flat, 150 mm wide 1 - HERA50073666
Supplier Article Number:: 50073666
Worksurface module, one-piece flat, 300 mm wide 1 - HERA50072685
Supplier Article Number:: 50072685
Worksurface module, One-piece flat, 600 mm wide 1 - HERA50073667
Supplier Article Number:: 50073667
Worksurface module, One-piece flat, 900 mm wide 1 - HERA50076398
Supplier Article Number:: 50076398
Worksurface module, One-piece indented, 600 mm wide 1 - HERA50073668
Supplier Article Number:: 50073668
Worksurface module, One-piece indented, 900 mm wide 1 - HERA50073669
Supplier Article Number:: 50073669
Worksurface module, One-piece indented, 1500 mm wide 1 - HERA50073671
Supplier Article Number:: 50073671
Arm rest AS3 prevents covering of ventilation openings hooks into ventilation slots 285mm wide Thermo Scientific Heraeus 1 - HERA50044976
Supplier Article Number:: 50044976
Footrest bar 0,9 m 1 - HERA50051982
Supplier Article Number:: 50051982
Footrest bar 1,2 m 1 - HERA50051983
Supplier Article Number:: 50051983
Footrest bar 1,5 m 1 - HERA50051984
Supplier Article Number:: 50051984
Footrest bar 1,8 m 1 - HERA50051985
Supplier Article Number:: 50051985
Electric adjustable floor stand 0,9 m with cable remote control, 750 to 950 mm working height 1 - HERA50117449
Supplier Article Number:: 50117449
Electric adjustable floor stand 1,2 m with cable remote control, 750 to 950 mm working height 1 - HERA50109314
Supplier Article Number:: 50109314
Electric adjustable floor stand 1,5 m with cable remote control, 750 to 950 mm working height 1 - HERA50117450
Supplier Article Number:: 50117450
Hanging bar with set of 6 hooks, for 0,9 m cabinet only 1 - HERA50073943
Supplier Article Number:: 50073943
Hanging bar with set of 12 hooks (may not be used with 0,9 m cabinet) 1 - HERA50073944
Supplier Article Number:: 50073944
Hose connection to fit through side wall access port, fits 10-13 mm diameter tubing 1 - HERA50059017
Supplier Article Number:: 50059017
Hose connection to fit through side wall access port, fits 3 mm diameter tubing 1 - HERA50066668
Supplier Article Number:: 50066668
Rear wall 3 service taps, left side of cabinet 1 - HERA51900649
Supplier Article Number:: 51900649
Rear wall 3 service taps, right side of cabinet 1 - HERA51900711
Supplier Article Number:: 51900711
Rear wall 6 service taps total, 3 on left and 3 on right side of cabinet, May not be used with 0,9m cabinet 1 - HERA51900722
Supplier Article Number:: 51900722
Drain pan valve 1 - HERA51900641
Supplier Article Number:: 51900641
Magnehelic pressure gauge 1 - HERA51900650
Supplier Article Number:: 51900650
2 Utility baskets including additional hanging bar 1 - HERA50078901
Supplier Article Number:: 50078901
Grid pre-filter for 1,5 and 1,8 m 5 - HERA50076921
Supplier Article Number:: 50076921
Vibration damped granite block 300 mm width. For weighing equipment and microscopy 1 - HERA50084339
Supplier Article Number:: 50084339
Battery backup for motorized front window 1 - HERA51900786
Supplier Article Number:: 51900786
Activated carbon pre-filter set 1 - HERA50083345
Supplier Article Number:: 50083345
Additional exhaust filter AEF 1 - HERA50047357
Supplier Article Number:: 50047357
Castors, for fixed height floor stands only (not to be used with electric stands) 1 - HERA50109977
Supplier Article Number:: 50109977
Exhaust air fittings with flow check flaps, for connections to exhaust system 1 - HERA50045033
Supplier Article Number:: 50045033
Hose connection to fit through side wall access port, fits 3 mm inner and 6 mm outer diameter tubing 1 - HERA50076408
Supplier Article Number:: 50076408
Intake screen GF 1 for HS 5 - HERA50047355
Supplier Article Number:: 50047355
Intake screen GF 3 for HS 5 - HERA50047356
Supplier Article Number:: 50047356
One-piece recessed work panel, AM 18 (1,8 m) 1 - HERA50044291
Supplier Article Number:: 50044291
One-piece recessed work panel, AM 6 (0,6 m) 1 - HERA50046087
Supplier Article Number:: 50046087
One-piece recessed work panel, AM 9 (0,9 m) 1 - HERA50044288
Supplier Article Number:: 50044288
One-piece work panel, AM 6 (0,6 m) 1 - HERA50045643
Supplier Article Number:: 50045643
One-piece work panel, AM 9 (0,9 m) 1 - HERA50045644
Supplier Article Number:: 50045644
Proper Use Pack,
prefilter for HSP, 12 mm
1 - HERA50047401
Supplier Article Number:: 50047401
Proper Use Pack,
prefilter for HSP, 15 mm
1 - HERA50047402
Supplier Article Number:: 50047402
Proper Use Pack,
prefilter for HSP, 18 mm
1 - HERA50047403
Supplier Article Number:: 50047403
Proper Use Pack,
prefilter for HSP, 9 mm
1 - HERA50047400
Supplier Article Number:: 50047400
Proper Use Pack, primary filter, 15 mm 1 - HERA50045647
Supplier Article Number:: 50045647
Proper Use Pack, primary filter, 18 mm 1 - HERA50045648
Supplier Article Number:: 50045648
Proper Use Pack, primary filter, 9 mm 1 - HERA50045645
Supplier Article Number:: 50045645
Proper Use Pack
activated charcoal
filter for HSPC, 12 mm
1 - HERA50051466
Supplier Article Number:: 50051466
Proper Use Pack
activated charcoal
filter for HSPC, 15 mm
1 - HERA50051467
Supplier Article Number:: 50051467
Proper Use Pack
activated charcoal
filter for HSPC, 18 mm
1 - HERA50051468
Supplier Article Number:: 50051468
Proper Use Pack
activated charcoal
filter for HSPC, 9 mm
1 - HERA50051465
Supplier Article Number:: 50051465
Spare holder for remote control 1 - HERA50075089
Supplier Article Number:: 50075089
Spare remote control set 1 - HERA50075090
Supplier Article Number:: 50075090
Worksurface module, one-piece flat, 1200 mm wide 1 - HERA50082970
Supplier Article Number:: 50082970
Worksurface module, one-piece flat, 1500 mm wide 1 - HERA50122210
Supplier Article Number:: 50122210
Worksurface module, one-piece flat, 1800 mm wide 1 - HERA50085281
Supplier Article Number:: 50085281
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty