Would you like to log in again?
Disposable particle filtering half masks, FFP1/FFP2/FFP3, K100 series
Supplier: 3M
The K100 Series Particulate Respirators provide effective respiratory protection for use in industries where workers will be exposed to dust particles and/or non-volatile liquid particles. The exhalation valve (models K111, K112 and K113) offers improved comfort in hot humid environments and/or where work is hard and physical. The filter and the valve are made of polypropylene and do not contain components made from natural rubber latex.
  • Flat design enables product to be easily stored when not in use
  • Diamond-shaped front allowd the respirator to maintain its shape in hot and humid environments
  • Single loop strap allowed easy adjustment on the face and head
  • Coloured noseclip for easy identification: yellow for FFP1, blue for FFP2 and red for FFP3
Certifications: EN 149:2001+A1:2009
ORDER
Expand All / Collapse All
Type Class With valve Size Pk VWR Catalog Number Quantity
K101 FFP1 No U* 20 MMMAK101 Unavailable
Supplier Article Number:: K101
K111 FFP1 Yes U* 10 MMMAK111 Unavailable
Supplier Article Number:: K111
K112 FFP2 Yes U* 10 MMMAK112 Unavailable
Supplier Article Number:: K112
K113 FFP3 Yes U* 5 MMMAK113 Unavailable
Supplier Article Number:: K113
unavailable This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +65 6505 0760.
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty