Trichloroethylene ≥97%, GPR RECTAPUR®

Danger

Synonyms: 1,1,2-TrichloroetheneTrichloroethene1,1,2-Trichloroethylene

VWRC28733.295EA 122 SGD
VWRC28733.295 VWRC28733.320
Trichloroethylene ≥97%, GPR RECTAPUR®
Trichloroethylene
Formula: ClCH=CCl₂
MW: 131,39 g/mol
Boiling Pt: 86,7 °C (1013 hPa)
Melting Pt: –73 °C
Density: 1,46 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000838
CAS Number: 79-01-6
EINECS: 201-167-4
UN: 1710
ADR: 6.1,III
Merck Index: 12,09769
https://sg.vwr.com/stibo/web/std.lang.all/13/81/741381.jpg

Order Now

Specification Test Results

Assay Min. 97 %
IR Spectrum Passes test
Density (20/4) 1.460 - 1.470
Evaporation residue Max. 50 ppm
Free alkali Max. 0.01 meq/g
n 20/D 1.460 - 1.480
Water Max. 0.02 %
Cl (Chloride) Max. 1 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR