Trichloroethylene ≥99.0%, AnalaR NORMAPUR® analytical reagent
|
Danger
|
Synonyms:
1,1,2-Trichloroethene
, Trichloroethene
, 1,1,2-Trichloroethylene
VWRC28735.292EA
133
SGD
VWRC28735.292
Trichloroethylene ≥99.0%, AnalaR NORMAPUR® analytical reagent
Trichloroethylene
Formula:
ClCH=CCl₂ MW: 131,39 g/mol Boiling Pt: 86,7 °C (1013 hPa) Melting Pt: –73 °C Density: 1,46 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000838 CAS Number: 79-01-6 EINECS: 201-167-4 UN: 1710 ADR: 6.1,III Merck Index: 12,09769 |
Specification Test Results
Assay (on anhydrous substance) | Min. 99.0 % |
Acidity | Max. 0.0001 meq/g |
Alkalinity | Max. 0.0006 meq/g |
Boiling point | 86.0 - 87.5 °C |
Colouration | Max. 10 APHA |
Density (20/4) | 1.460 - 1.470 |
Refractive index (20°C) | 1.460 - 1.480 |
Evaporation residue | Max. 50 ppm |
Free halogens | Max. 0.3 ppm |
Water | Max. 100 ppm |
Al (Aluminium) | Max. 0.5 ppm |
B (Boron) | Max. 0.02 ppm |
Ba (Barium) | Max. 0.1 ppm |
Ca (Calcium) | Max. 0.5 ppm |
Cd (Cadmium) | Max. 0.05 ppm |
Co (Cobalt) | Max. 0.02 ppm |
Cr (Chromium) | Max. 0.02 ppm |
Cu (Copper) | Max. 0.02 ppm |
Fe (Iron) | Max. 0.1 ppm |
Mg (Magnesium) | Max. 0.1 ppm |
Mn (Manganese) | Max. 0.02 ppm |
Ni (Nickel) | Max. 0.02 ppm |
Pb (Lead) | Max. 0.1 ppm |
Sn (Tin) | Max. 0.5 ppm |
Zn (Zinc) | Max. 0.1 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...