2-Chloro-4-nitrobenzoic acid ≥98%

Supplier: Thermo Scientific
Danger

ALFAA11602.36EA 0 SGD
ALFAA11602.36
2-Chloro-4-nitrobenzoic acid ≥98%
2-Chloro-4-nitrobenzoic acid

Purity: 98%
CAS No.: 99-60-5
Molecular Formula: C7H4ClNO4
Formula Weight: 201.57
Odour: Odourless
Melting Point: 140-144°
Beilstein Reference: 1912836
MDL No.: MFCD00007209

Formula: C₇H₄ClNO₄
MW: 201,57 g/mol
Melting Pt: 140…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00007209
CAS Number: 99-60-5
EINECS: 202-771-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR