Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,3,5-Trifluorobenzene ≥98%

Supplier: Alfa Aesar

Purity: 98+%
CAS No.: 372-38-3
Molecular Formula: C6H3F3
Formula Weight: 132.09
Flash Point: -7°(19°F)
Refractive Index: 1.4150
Melting Point: -7 to -4°
Boiling Point: 75-76°
Beilstein Reference: 1906457
UN No.: 2387
MDL No.: MFCD00000333

Formula: C₆H₃F₃
MW: 132,09 g/mol
Boiling Pt: 75 °C (1013 hPa)
Density: 1,277 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000333
CAS Number: 372-38-3
EINECS: 206-751-2
UN: 2387
ADR: 3,II


Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 5 g ALFAA15984.06
Supplier Article Number:: A15984.06
Glass bottle for solids 100 g ALFAA15984.22
Supplier Article Number:: A15984.22
Glass bottle for solids 25 g ALFAA15984.14
Supplier Article Number:: A15984.14
Expand All / Collapse All

SPECIFICATIONS

Melting Point -7 to -4°
Formula Weight 132.09
CAS No. 372-38-3
Purity 98+%
Refractive Index 1.4150
Flash Point -7°(19°F)
Boiling Point 75-76°
Molecular Formula C6H3F3
MDL No. MFCD00000333
UN No. 2387
Beilstein Reference 1906457
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty