ALFAA12380

ALFAA12380.14EA 70.1 SGD
ALFAA12380.14 ALFAA12380.22
ALFAA12380
Purity: 98%
CAS No.: 321-28-8
Molecular Formula: C7H7FO
Formula Weight: 126.13
Flash Point: 56°(133°F)
Refractive Index: 1.4940
Melting Point: -39°
Boiling Point: 154-155°
Beilstein Reference: 1859755
UN No.: 1993
MDL No.: MFCD00000316
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR